Honeywell™ Uvex™ Genesis™ Protective Safety Glasses
Three-position ratchet lens inclination system and adjustable temples enable wearer to customize fit
Supplier:  Honeywell™ UVS3242
 View more versions of this product
Catalog No. 19-040-172
Explore available promotions
Description
Description
  • Impact-resistant polycarbonate lenses block >99.9% of harmful UVA and UVB radiation
  • Ventilation channel between frame and lens prevents moisture buildup
  • Nose bridge is covered with small, ultrasoft “fingers” that hug the nose for improved fit and comfort
  • Soft elastomer browguard cushions all points of contact with face
uvextreme™ Anti-Fog Coating (XTR)
  • Permanently bonded to lens
  • Absorbs 99.9% of UV radiation
  • Anti-scratch and -static
  • Will not wear after repeated cleanings
Hydroshield™ Anti-Fog Coating
  • Provides 99.9% ultra-violet protection
  • Provides 2X greater scratch-resistance
XC Series
  • Extended wrap-around lens for extra cheek and extra side protection
    • Soft flexible nose fingers conform to fit a variety of nasal profiles
    • Adjustable temples and lens inclination
    ultra-dura™ Hard Coat (UD)
    • Permanently bonded to lens
    • Scratch- and chemical-resistant
    Compliance
    Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards. Meet stringent military V0 Ballistic Test for Impact requirements.
    Warranty
    Lifetime frame guarantee.
Specifications
Specifications
Meets ANSI Z87+ and CSA Z94.3 standards
Lifetime frame guarantee
Amber
Vapor Blue
SDS
Documents
Documents
Accessories