Honeywell™ Uvex™ Stealth™ OTG Goggles
Low-profile, wraparound style provides complete coverage designed for all-day wear
Supplier:  Honeywell™ S3970HSF
 View more versions of this product
Catalog No. 296367471
Explore available promotions
Description
Description
  • Ideal for impact, dust/airborne particles, chemical splash and optical radiation (UV and welding/heat radiation)
  • Superior optics and panoramic field of vision enhanced with toric lens
  • Pivoting clip and quick-adjusting wide headband
  • Elastomer body conforms and contours for a secure fit—even over most prescription eyewear
  • Replacement lenses easily snap on
Specifications
Specifications
ANSI Z87.1-2015, CSA Z94.3
OTG Goggles
Navy
Clear
Ideal for most indoor work applications
Fabric