Honeywell™ Uvex™ XC™ Safety Glasses Replacement Lenses
Extended wrap-around lens for extra cheek and extra side protection. Soft flexible nose fingers conform to fit a variety of nasal profiles. Adjustable temples and lens inclination.
$78.79 - $104.91
Specifications
For Use With (Equipment) XC Spectacles
View More Specs
Products 2
Catalog Number Mfr. No. Lens Tint Price Quantity  
Catalog Number Mfr. No. Lens Tint Price Quantity  
19-066-043
View Documents
Honeywell™
UVS6950
Clear
Pack of 10 Each for $78.79
19-066-047
View Documents
Honeywell™
UVS6952
SCT-Reflect 50
Pack of 10 Each for $104.91
Description
Description
Uvex Genesis XC has the same comfort and adjustability features as original Genesis, with 20% more extended wrap-around lens for extra cheek and side coverage. Rx lens insert available. Uvex Genesis XC has an easy and economical lens replacement system.
  • PC lens, Hydroshield AF, Dura-streme AF/HC
  • Available in a very wide range of lens tints
  • 20% more coverage than original Genesis
  • Economical lens replacement system
  • Meets military Vo ballistic impact requirement (on select tints)
Specifications
Specifications
XC Spectacles
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'