UVP Camera Filter SYBR Green
Allows visualization of fluorophores from 460-470nm
Supplier:  UVP 38021901
Catalog No. UVP38021901
Description
Description
  • Blocks UV and IR radiation emitted from the transilluminator
Specifications
Specifications
50
For use with chromadoc imaging system
Camera Filter Sybr Green