UVP DigiDoc-It™ Imaging Systems
Economical gel imaging systems with digital color camera, compact hood, high-performance transilluminator, capture software and sample viewer, captures high quality gel images
Supplier:  UVP 97024601
 View more versions of this product
Catalog No. UV97024601
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Includes: Digital camera, with 55mm Ethidium bromide filter, USB interface; Doc-It Capture Software on CD-ROM; Hood enclosure; High-performance 25-watt UV transilluminator (UVP97010501 is compatible with a variety of UVP enchtop transilluminators)
Description
Description
  • Camera-hood system provides low cost image acquisition and documentation of gels, plates and membranes
  • Combines high megapixel digital color camera and compact hood enclosure
  • Hood with viewport fits on top of UVP's High Performance UV transilluminator
  • Uses your existing computer with Doc-It™ capture software on supplied CD
  • High-quality images with low-light capability
  • Software enables image capture, complete camera control, enhancement, annotation and archiving of digital images for publication or quantitative analysis
  • Model without transilluminator is compatible with optional Drawer (UVP98006801)and Benchtop UV Transilluminators
Camera and Capture Software
  • Color camera with high resolution
  • High or low image compression with multiple file formats:JPEG, BMP, TIF, GIF, TGA, PNG
  • Capture software is compatible with Windows™ XP (SP2) and newer
Hood
  • Lightweight hood with viewport and easy access door Measures 12D × 173/4W × 165/8 in.H (305 × 451 × 422mm)
Specifications
Specifications
25 x 20 cm
High-performance TFM-30V
Imaging System
Digital camera, with 55mm Ethidium bromide filter, USB interface; Doc-It Capture Software on CD-ROM; Hood enclosure; High-performance 25-watt UV transilluminator (UVP97010501 is compatible with a variety of UVP enchtop transilluminators)
10MP Digital
Product Suggestions
Product Suggestions