UVP EL Series 2UV Lamps
Select from lamps with four, six or eight watts
Supplier:  UVP 95025101
 View more versions of this product
Catalog No. UVP95025101
Explore available promotions
Description
Description
  • Manufactured with a durable, extruded metal housing and scratch resistant powder paint
  • Lightweight and easy to handle
  • Include two different UV wavelengths in one lamp
  • Can be placed in the J-138 stand for hands-free use
Applications:
Bacterial identification, specimen staining, gel electrophoresis, optical lab measurements, fluorescence demonstration, biochemical testing
Specifications
Specifications
Handheld UV Lamp
115 V
365/302 nm
Metal
64 mm
8 W, 115 V, 60 Hz
60 Hz
376 mm
1 Ea.
97 mm
UV-A, UV-B
8 W
0.16 A
2.5 in.
14.8 x 3.8 in. (376 x 97 mm)
Bacterial identification, specimen staining, Gel electrophoresis, optical lab measurements, fluorescence demonstration, biochemical testing
14.8 in.
UVLS-28 Lamp
3.8 in.