UVP Mini Ultraviolet Lamp Replacement Tube
Replacement tubes for lightweight, compact and financial UV Lamps
Supplier:  UVP 34000401
 View more versions of this product
Catalog No. UVP34000401
Explore available promotions
Description
Description
Specifications
Specifications
4 W
Model UVGL-15 and UVSL-14P compact and mini handheld UV lamp
134.9 mm
365/312 nm
5.31 in.
1 Ea.