missing translation for 'onlineSavingsMsg'
Learn More

UVP Pen-Ray™ Lamp Power Supplies

Provide power to Pen-Ray lamps. Electrical requirements, 115V 50/60Hz. ETL/ETLC certified.

Supplier:  UVP 99005501

Catalog No. UVP99005501


Only null left
Explore available promotions

Description

Description

  • Provide power to Pen-Ray lamp models 11SC-1, 11SC-2, 11SC1-L, 3SC-9
  • Electrical requirements, 115V 50/60Hz

Certifications

ETL/ETLC certified

Specifications

Specifications

18 A
Pen-Ray Mercury Lamp, Rare Gas Lamp
58 mm
6.5 in.
1 Ea.
94 mm
6.5 x 3.7 x 2.3 in. (165 x 94 x 58 mm)
2.3 in.
60 Hz
165 mm
3.7 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions