UVP Replacement UV Tubes
For use with 8w UV lamps.
Supplier:  UVP 34000601
 View more versions of this product
Catalog No. UVP34000601
Explore available promotions
Description
Description
For use with 8w UV lamps.
  • For use with 8w UV lamps
Specifications
Specifications
Replacement UV Tube
365 nm
11.33 in.
8 W
Model TLW-20 transilluminator, model CL-1000 crosslinker, models 2UV and 3UV transilluminator and EL series UV lamp
28.7 cm