UVP Replacement UV Tubes
For use with 8w UV lamps.
Supplier:  UVP 34004201
 View more versions of this product
Catalog No. UVP34004201
Explore available promotions
Description
Description
For use with 8w UV lamps.
  • For use with 8w UV lamps
Specifications
Specifications
Replacement UV Tube
8 W
302 nm
CL-1000M crosslinker, TMW-20 and benchtop series transilluminator and EL series UV lamp