missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Greiner Bio-One VACUETTE™ EVOPROTECT SAFETY Blood Collection Set and Luer Adapter

Blood collection set with luer adapter is not made with natural rubber latex or DEHP

Supplier:  Greiner Bio-One 450127

 View more versions of this product

Catalog No. 22-405-172

Explore available promotions

Description

Description

  • Blood collection set
  • Luer adapter
  • Single packed
  • Sterile
  • 0.75 in. tubing
Specifications

Specifications

Safety Blood Collection Set with Luer Adapter
21 G
Blood collection
7.5 in.
Green
Sterile
Luer adapter
60 Pk.
Videos
SDS
Documents

Documents

Promotions

Promotions