BD Vacutainer™ Blood Transfer Device
Blood Transfer Device meets all procedural and safety standards for blood transfer from a conventional hypodermic needle and syringe to a blood collection tube or blood culture bottle.
Supplier: BD 36488000
Description
- The use of a needle to transfer venous blood from a syringe to a blood collection tube or blood culture bottle is both a dangerous procedure and an OSHA-prohibited practice.
- The BD Vacutainer™ blood transfer device was designed with healthcare workers’ safety in mind.
- The single-use device reduces the risk of transfer-related injuries, while maintaining the specimen integrity required for accurate results.
Specifications
Non-sterile | |
Blood Transfer Device | |
Plastic |