missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Vacutainer™ Plastic Urinalysis Tubes
For the collection and transport of urine specimens. BD Vacutainer™ Plastic Urinalysis Tubes are sterile evacuated tubes with or without preservative to be used with BD collection devices. Available in many sizes with a round or conical bottom.
$101.79 - $101.79
Specifications
Product Type | Urinalysis Tube |
---|---|
Disposable | Single-use |
Material | PET |
Sterility | Sterile |
For Use With (Application) | Urinalysis |
Catalog Number | Mfr. No. | Volume (Metric) | Bottom Shape | Closure Type | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Bottom Shape | Closure Type | Price | Quantity | |||
22-204-371
|
BD
364992 |
8 mL | Conical | Conventional Rubber Stopper |
Pack of 100 Each for $101.79
Case of 10 Pack for $1,017.90
|
|
|||
Description
BD Vacutainer™ urine products for the collection and transport of urine specimens offer the same level of safety and standardization as BD blood collection products.- Designed to provide accurate patient results with reduced risk of healthcare worker exposure to hazardous specimens
- Used with BD collection devices, specimen cups, 24 hour 3L containers and transfer straws
- Leak-proof urine tubes can safely transport urine specimens to the laboratory for analysis
- Options include bottom shape (round or conical) and cap style (conventional or Hemogard™)
- Most sizes are preservative/additive free
- 8mL tube (mfr. no. 364938) contains a preservative: 0.4% Chlorhexidine, 5.6% Ethyl Paraben and 94% Sodium Propionate
Specifications
Urinalysis Tube | |
PET | |
Urinalysis | |
Paper |
Single-use | |
Sterile | |
Urine collection devices (cups, transfer straws, 24 hr. containers) |