DWK Life Sciences Kimble™ Vacuum Desiccator with Collar 200x310
Ideal for removing moisture from solids, storage of hygroscopic chemicals, and applications requiring vacuum desiccation.
Supplier:  DWK Life Sciences 31200200
Catalog No. 12-141-434
Explore available promotions
Description
Description
  • Greased rotating collar valve is used to facilitate vacuum desiccation
  • Includes solid glass lid with collar valve, metal plate, and a recessed base for desiccant containment
  • Manufactured from soda-lime glass conforming to USP Type III and ASTM E438, Type II requirements
  • Can be used for cooling oven-dried samples while maintaining sample dryness
Specifications
Specifications
Vacuum Desiccator with Collar