missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Valeraldehyde 95.0+%, TCI America™
$44.79 - $145.98
Chemical Identifiers
CAS | 110-62-3 |
---|---|
Molecular Formula | C5H10O |
Molecular Weight (g/mol) | 86.13 |
MDL Number | MFCD00007026 |
InChI Key | HGBOYTHUEUWSSQ-UHFFFAOYSA-N |
Synonym | valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde |
PubChem CID | 8063 |
ChEBI | CHEBI:84069 |
IUPAC Name | pentanal |
SMILES | CCCCC=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
V000125ML
|
TCI America
V000125ML |
25 mL |
Each for $44.79
|
|
|||||
V0001500ML
|
TCI America
V0001500ML |
500 mL |
Each for $145.98
|
|
|||||
Chemical Identifiers
110-62-3 | |
86.13 | |
HGBOYTHUEUWSSQ-UHFFFAOYSA-N | |
8063 | |
pentanal |
C5H10O | |
MFCD00007026 | |
valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde | |
CHEBI:84069 | |
CCCCC=O |
Specifications
110-62-3 | |
102°C | |
MFCD00007026 | |
2058 | |
HGBOYTHUEUWSSQ-UHFFFAOYSA-N | |
pentanal | |
8063 | |
86.13 | |
Liquid |
Yellow | |
C5H10O | |
25 mL | |
valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde | |
CCCCC=O | |
86.13 | |
CHEBI:84069 | |
≥95.0% (GC) | |
Valeraldehyde |
Safety and Handling
EINECSNumber : (2)-0488&(2)-0494
RTECSNumber : YV3600000
TSCA : Yes