missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Valeric acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A16238AE

 View more versions of this product

Catalog No. AAA16238AE


Explore available promotions

Description

Description

Valeric acid, is used as a sex attractant of the sugar beet wireworm, Limonius californicus. It is used predominantly as an intermediate in the manufacture of flavors and perfumes, ester type lubricants, plasticizers and vinyl stabilizesrs. It is a food additive used as a synthetic flavoring substance dan adjuvant. It is an inert ingredient in controlling pest.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valeric acid, is used as a sex attractant of the sugar beet wireworm, Limonius californicus. It is used predominantly as an intermediate in the manufacture of flavors and perfumes, ester type lubricants, plasticizers and vinyl stabilizesrs. It is a food additive used as a synthetic flavoring substance dan adjuvant. It is an inert ingredient in controlling pest.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

Valeric acid
109-52-4
C5H10O2
MFCD00004413
969454
valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n
NQPDZGIKBAWPEJ-UHFFFAOYSA-N
pentanoic acid
7991
102.13
0.938
88°C (190°F)
1.408
-34°C
184°C to 186°C
CH3(CH2)3CO2H
UN3265
14,9904
It is soluble in water.
CCCCC(=O)O
102.133
CHEBI:17418
99%
100 mL
Unpleasant

Chemical Identifiers

109-52-4
102.133
NQPDZGIKBAWPEJ-UHFFFAOYSA-N
7991
pentanoic acid
C5H10O2
MFCD00004413
valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n
CHEBI:17418
CCCCC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H314-H227
Causes severe skin burns and eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 203-677-2

RTECSNumber : YV6100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only