missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Value Combo Refrigerator/Freezer

Chill, freeze and retrieve samples safely, simply and economically with the Fisherbrand™ Value Refrigerator/Freezer.

Supplier:  Fisherbrand™ FBV18CPSA

Catalog No. FBV18CPSA


Explore available promotions

Description

Description

  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally-friendly, SNAP-compliant hydrocarbon refrigerants
  • Adjustable temperature control
  • Easy to clean, durable finish
  • Manual and auto-defrost combination units
Temperature Range: 
  • Refrigerator: 1°C to 12°C (33.8°F to 53.6°F)
  • Freezer: -24°C to -12°C (-11.2°F to +10.4°F)
Specifications

Specifications

Combination General Purpose Refrigerator/Freezer
510 L
Adjustable
Hinged
Right
3
Cyclopentane
cULus
White
Indoor general storage of non-critical samples and reagents
Automatic
4 in. on Sides, 4 in. at Back, 11 in. on Top
Refrigerator: 64.7 x 63.5 x 101.6 cm (25.5 x 25 x 40 in.); Freezer: 52 x 57.1 x 40 cm (20.5 x 22.5 x 15.75 in.)
1.1 A
115 V
50 Hz
101.6 kg
18 cu. ft.
Refrig: +1°C to +12°C (+33.8°F to +53.6°F); Freezer: -24°C to -12°C (-11.2°F to +10.4°F)
R600a
2 (2 compartments)
2 Adjustable Shelves, 2 Drawers (Refrigerator); 1 Shelf (Freezer)
Enameled Steel
ABS Plastic
2 adjustable leveling feet
Electronic Controls
10 years
Interior
Bottom-mounted
83.3 x 75 x 169.2 cm (32.79 x 29.53 x 66.61 in.)
NEMA 5-15 P
115 V/60 Hz
224 lb.
Videos
SDS
Documents

Documents

Promotions

Promotions