DWK Life Sciences Kimble™ ValueWare™ Low-Form Griffin Beakers
With spout.
$106.05 - $146.95
Specifications
Material | Borosilicate Glass |
---|---|
For Use With (Application) | Measuring and Mixing Liquids Where a High Level of Accuracy is Not Required |
Includes | Spout |
Label Type | Matte Finish Marking |
Type | Beaker |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
S00001
|
DWK Life Sciences
5400010 |
10 mL |
Pack of 12 Each for $121.56
Case of 4 Pack for $486.24
|
|
|||||
S00002
|
DWK Life Sciences
5400020 |
20 mL |
Pack of 12 Each for $119.85
Case of 4 Pack for $479.40
|
|
|||||
S00005
|
DWK Life Sciences
5400050/EMD |
50 mL |
Pack of 12 Each for $122.36
Case of 4 Pack for $489.44
|
|
|||||
S00006
|
DWK Life Sciences
54000100 |
100 mL |
Pack of 12 Each for $106.05
Case of 4 Pack for $424.20
|
|
|||||
S00008
|
DWK Life Sciences
54000250 |
250 mL |
Pack of 12 Each for $111.74
Case of 4 Pack for $446.96
|
|
|||||
S00012
|
DWK Life Sciences
540001000 |
1000 mL |
Pack of 6 Each for $146.95
Case of 4 Pack for $587.80
|
|
|||||
Description
- Thick, slightly flared, beaded top
- With spout designed to have excellent pouring characteristics
- Uniform Kimax™ KG-33 glass walls throughout
- Durable matte-finish marking area for use with an ordinary pencil
- Easy-to-read white graduated scale on 0.67 to 33.8 oz. (20 to 1000mL) sizes for measuring and/or mixing liquids where a high level of accuracy is not required
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Beaker designed to ASTM E-960, Type I specification requirements.
Specifications
Borosilicate Glass | |
Spout | |
Beaker | |
Griffin |
Measuring and Mixing Liquids Where a High Level of Accuracy is Not Required | |
Matte Finish Marking | |
ASTM E438, ASTM E960 |