DWK Life Sciences Kimble™ ValueWare™ Test Tubes
Made from Kimax™ KG-33 glass tubing.
$58.67 - $58.67
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) | Length (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) | Length (Metric) | Price | Quantity | |||
S00177
|
DWK Life Sciences
550421275 |
6 mL | 12 mm | 75 mm |
Pack of 72 Each for $58.67
Case of 10 Pack for $586.70
|
|
|||
Description
- Sturdy lip
- Uniform wall thickness for maximum heat transfer and chemical resistance
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E982, Type I specification requirements.
Specifications
Borosilicate Glass |