missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

Vanadium foil, 0.25mm (0.01in) thick, 99.5% (metals basis), Thermo Scientific Chemicals

Form: approx. 13g/50x150mm

$392.63 - $1269.19

Identifiants chimiques

CAS 7440-62-2
Molecular Formula V
Molecular Weight (g/mol) 50.94
MDL Number MFCD00011453
InChI Key LEONUFNNVUYDNQ-UHFFFAOYSA-N
Synonym dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
PubChem CID 23990
ChEBI CHEBI:35170
IUPAC Name vanadium
SMILES [V]
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AA14633FZ
Afficher les documents
Thermo Scientific Chemicals
014633FZ
50 x 150 mm
chaque for $392.63
 
AA14633GU
Afficher les documents
Thermo Scientific Chemicals
014633GU
150 x 200 mm
chaque for $1,269.19
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

7440-62-2
50.94
LEONUFNNVUYDNQ-UHFFFAOYSA-N
23990
vanadium
V
MFCD00011453
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
CHEBI:35170
[V]

Spécifications

Silver to Gray
Silver Gray foil
V
50 x 150 mm
LEONUFNNVUYDNQ-UHFFFAOYSA-N
vanadium
23990
Odorless
Vanadium
7440-62-2
99.5% (Metals basis)
MFCD00011453
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
[V]
50.94
CHEBI:35170
Foil
Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : 231-171-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only