missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanadium(III) fluoride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 08111504

 View more versions of this product

Catalog No. AA8111504


Explore available promotions

Description

Description

Employed in oxygen-sensitive applications, such as metal production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Vanadium (III) fluoride
10049-12-4
2 g
MFCD00042553
14,9922
Practically insoluble in water, acetone, and glacial acetic acid
[F-].[F-].[F-].[V+3]
107.94
98%
3.363 g/mL at 19°C
≥1406°C
Powder
F3V
UN2923
Vanadium trifluoride
PASVSMBJEYQYSN-UHFFFAOYSA-K
vanadium(3+) trifluoride
107.94
Moisture sensitive

Chemical Identifiers

10049-12-4
107.94
PASVSMBJEYQYSN-UHFFFAOYSA-K
vanadium(3+) trifluoride
F3V
MFCD00042553
Vanadium trifluoride
[F-].[F-].[F-].[V+3]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H314-H318
Fatal if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301-H314-H330-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

EINECSNumber : 233-169-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only