missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Vanadium(IV) oxide meso-tetraphenylporphine, Thermo Scientific Chemicals
$339.67 - $339.67
Chemical Identifiers
CAS | 14705-63-6 |
---|---|
Molecular Formula | C44H30N4O2V |
Molecular Weight (g/mol) | 697.692 |
MDL Number | MFCD00012154 |
InChI Key | LRQXAIDMLNPKOR-UHFFFAOYSA-N |
Synonym | 5,10,15,20-tetraphenyl-21h,23h-porphine vanadium iv oxide |
PubChem CID | 131699123 |
IUPAC Name | oxygen(2-);5,10,15,20-tetraphenyl-21,22-dihydroporphyrin;vanadium(4+) |
SMILES | C1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9)N3.[O-2].[O-2].[V+4] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AA39583ME
|
Thermo Scientific Chemicals
039583ME |
500 mg |
Each for $339.67
|
|
||||||
Description
It is used for research in catalysis. Thin films of VOTPP may be used in detecting methane gas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14705-63-6 | |
697.692 | |
LRQXAIDMLNPKOR-UHFFFAOYSA-N | |
131699123 | |
C1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9)N3.[O-2].[O-2].[V+4] |
C44H30N4O2V | |
MFCD00012154 | |
5,10,15,20-tetraphenyl-21h,23h-porphine vanadium iv oxide | |
oxygen(2-);5,10,15,20-tetraphenyl-21,22-dihydroporphyrin;vanadium(4+) |
Specifications
Purple | |
Crystalline | |
C44H30N4O2V | |
5,10,15,20-tetraphenyl-21h,23h-porphine vanadium iv oxide | |
LRQXAIDMLNPKOR-UHFFFAOYSA-N | |
oxygen(2-);5,10,15,20-tetraphenyl-21,22-dihydroporphyrin;vanadium(4+) | |
131699123 | |
Odorless |
14705-63-6 | |
500 mg | |
MFCD00012154 | |
Insoluble in water. | |
C1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9)N3.[O-2].[O-2].[V+4] | |
697.692 | |
679.67 | |
Vanadium(IV) oxide meso-tetraphenylporphine |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only