missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Vanadium rod, 6.35mm (0.25in) dia, 99.5% (metals basis), Thermo Scientific Chemicals
$281.23 - $3707.14
Chemical Identifiers
CAS | 7440-62-2 |
---|---|
Molecular Formula | V |
Molecular Weight (g/mol) | 50.94 |
MDL Number | MFCD00011453 |
InChI Key | LEONUFNNVUYDNQ-UHFFFAOYSA-N |
Synonym | dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde |
PubChem CID | 23990 |
ChEBI | CHEBI:35170 |
IUPAC Name | vanadium |
SMILES | [V] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA13996BQ
|
Thermo Scientific Chemicals
013996BQ |
5 cm |
Each for $281.23
|
|
|||||
AA13996BU
|
Thermo Scientific Chemicals
013996BU |
25 cm |
Each for $1,011.70
|
|
|||||
AA13996BY
|
Thermo Scientific Chemicals
013996BY |
100 cm |
Each for $3,707.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-62-2 | |
50.94 | |
LEONUFNNVUYDNQ-UHFFFAOYSA-N | |
23990 | |
vanadium |
V | |
MFCD00011453 | |
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde | |
CHEBI:35170 | |
[V] |
Specifications
7440-62-2 | |
MFCD00011453 | |
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde | |
[V] | |
50.94 | |
CHEBI:35170 | |
Odorless | |
Rod | |
(metals basis) |
V | |
5 cm | |
LEONUFNNVUYDNQ-UHFFFAOYSA-N | |
vanadium | |
23990 | |
99.5% | |
≈1.93g/cm | |
Vanadium rod, 6.35mm (0.25 in.) dia. |
Safety and Handling
EINECSNumber : 231-171-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only