missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanadium(V) oxide, 98+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 207990025

Catalog No. AC207990025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Vanadium(V) oxide
1314-62-1
98% min
V2O5
15, 10107
Solubility in water: 8g/L. Other solubilities: soluble in acids and alkalies,insoluble in alcohol
O=[V](=O)O[V](=O)=O
181.88
CHEBI:30045
98+%
3.35
98+%
2.5kg
O5V2
MFCD00011457
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride
GNTDGMZSJNCJKK-UHFFFAOYSA-N
dioxovanadiooxy(dioxo)vanadium
14814
181.88
Plastic bottle
3.3500g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of damaging the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes damage to organs through prolonged or re

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

GHS Signal Word: Danger

EINECSNumber : 215-239-8

RTECSNumber : YW2450000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only