Learn More
Vanadyl(IV) acetylacetonate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 199590010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Vanadyl(IV) acetylacetonate | |
3153-26-2 | |
Authentic | |
Glass bottle | |
(CH3COCH=C(CH3)O)2V=O | |
1kg | |
vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide | |
DKHLKZMCKOJMTD-SUKNRPLKSA-L | |
(Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate | |
131674261 | |
99% |
99% | |
79°C | |
98.5% min | |
C10H14O5V | |
MFCD00000032 | |
02,456; 03,331; 11,47; 13,54; 15,57; 15,186 | |
Solubility in water: negligible. Other solubilities: soluble in methanol (6.4 g/100 mL), ethanol, ether, chloroform, acetone, dioxane, benzene (0.9 g/100 m, l), pyridine | |
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].O.[V+2] | |
265.15 | |
265.15 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
If eye irritation persists: Get medical
GHS Signal Word: Warning
EINECSNumber : 221-590-8
TSCA : TSCA
RUO â Research Use Only