Learn More
Vanadyl(IV) acetylacetonate, 99%, Thermo Scientific Chemicals
$147.32 - $147.32
Chemical Identifiers
CAS | 3153-26-2 |
---|---|
Molecular Formula | C10H14O5V |
Molecular Weight (g/mol) | 265.16 |
MDL Number | MFCD00000032 |
InChI Key | JFHJZWAQYMGNBE-SUKNRPLKSA-L |
Synonym | vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide |
PubChem CID | 131674261 |
IUPAC Name | (Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate |
SMILES | O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC199590500
|
Thermo Scientific Chemicals
199590500 |
50 g | Glass bottle |
Each for $147.32
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3153-26-2 | |
265.16 | |
JFHJZWAQYMGNBE-SUKNRPLKSA-L | |
131674261 | |
O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
C10H14O5V | |
MFCD00000032 | |
vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide | |
(Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate |
Specifications
3153-26-2 | |
Blue-Green to Green | |
Authentic | |
Glass bottle | |
(CH3COCH=C(CH3)O)2V=O | |
50 g | |
vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide | |
JFHJZWAQYMGNBE-SUKNRPLKSA-L | |
(Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate | |
131674261 | |
99% | |
Vanadyl(IV) acetylacetonate, 99% |
256.0°C to 259.0°C | |
79°C | |
98.5% min | |
C10H14O5V | |
MFCD00000032 | |
02,456; 03,331; 11,47; 13,54; 15,57; 15,186 | |
Solubility in water: negligible. Other solubilities: soluble in methanol (6.4 g/100 mL), ethanol, ether, chloroform, acetone, dioxane, benzene (0.9 g/100 m, l), pyridine | |
O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
265.16 | |
265.15 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
If eye irritation persists: Get medical
GHS Signal Word: Warning
EINECSNumber : 221-590-8
TSCA : TSCA
RUO – Research Use Only