Learn More
Vanadyl(IV) sulfate hydrate, 17-23% V, Thermo Scientific Chemicals
$197.44 - $367.55
Chemical Identifiers
CAS | 12439-96-2 |
---|---|
Molecular Formula | O5SV·xH2O |
Molecular Weight (g/mol) | 163 |
MDL Number | MFCD00149933 |
InChI Key | YGYBXHQARYQUAY-UHFFFAOYSA-L |
Synonym | oxosulfatovanadium pentahydrate, vanadyl sulfate pentahydrate, vanadium, oxosulfato-, pentahydrate, oxovanadium iv sulfate pentahydrate, vanadium 2+ , pentaaquaoxo-, oc-6-21 , sulfate 1:1, oxovanadium iv pentahydrate sulfate, vanadic sulfate pentahydrate, vanadyl sulphate pentahydrate, voso4.5h2o, ov.so4.5h2o |
PubChem CID | 167150 |
IUPAC Name | oxovanadium(2+);sulfate;pentahydrate |
SMILES | O.O.O.O.O.[O-]S(=O)(=O)[O-].O=[V+2] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC214840250
|
Thermo Scientific Chemicals
214840250 |
25 g | Glass bottle |
Each for $197.44
|
|
|||||
AC214841000
|
Thermo Scientific Chemicals
214841000 |
100 g | Glass Bottle |
Each for $367.55
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12439-96-2 | |
163 | |
YGYBXHQARYQUAY-UHFFFAOYSA-L | |
167150 | |
O.O.O.O.O.[O-]S(=O)(=O)[O-].O=[V+2] |
O5SV·xH2O | |
MFCD00149933 | |
oxosulfatovanadium pentahydrate, vanadyl sulfate pentahydrate, vanadium, oxosulfato-, pentahydrate, oxovanadium iv sulfate pentahydrate, vanadium 2+ , pentaaquaoxo-, oc-6-21 , sulfate 1:1, oxovanadium iv pentahydrate sulfate, vanadic sulfate pentahydrate, vanadyl sulphate pentahydrate, voso4.5h2o, ov.so4.5h2o | |
oxovanadium(2+);sulfate;pentahydrate |
Specifications
White to Yellow | |
Powder | |
O5SV·xH2O | |
MFCD00149933 | |
Solubility in water: soluble | |
O.O.O.O.O.[O-]S(=O)(=O)[O-].O=[V+2] | |
163 | |
163 | |
Glass bottle |
12439-96-2 | |
25 g | |
VOSO4·xH2O | |
oxosulfatovanadium pentahydrate, vanadyl sulfate pentahydrate, vanadium, oxosulfato-, pentahydrate, oxovanadium iv sulfate pentahydrate, vanadium 2+ , pentaaquaoxo-, oc-6-21 , sulfate 1:1, oxovanadium iv pentahydrate sulfate, vanadic sulfate pentahydrate, vanadyl sulphate pentahydrate, voso4.5h2o, ov.so4.5h2o | |
YGYBXHQARYQUAY-UHFFFAOYSA-L | |
oxovanadium(2+);sulfate;pentahydrate | |
167150 | |
17 to 23% (V) | |
Vanadyl(IV) sulfate hydrate, 17-23% V |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RTECSNumber : YW1840000
RUO – Research Use Only