missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cole-Parmer™ VapLock™ Solvent Waste Kits for Justrite™ Containers

VapLock™ Solvent Waste Kits for Justrite™ Containers collect effluent from multiple sources while minimizing harmful solvent exhaust

$1,044.00 - $1,256.03

Specifications

Closure Size 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F)
Size Justrite Quick-Disconnect
For Use With (Application) Solvent Safety
For Use With (Equipment) Solvent-Compatible Containers
View More Specs

Products 3
Catalog Number Mfr. No. Description Material Price Quantity  
Catalog Number Mfr. No. Description Material Price Quantity  
13-311-154
View Documents
Cole-Parmer™
1201952
Solvent Waste Kit, Polypropylene, Justrite Polypropylene Polypropylene N/A
 
13-311-155
View Documents
Cole-Parmer™
1201953
Solvent Waste Kit, Polypropylene, Justrite SS Polypropylene N/A
 
13-311-156
View Documents
Cole-Parmer™
1201954
Solvent Waste Kit, PTFE, Justrite SS PTFE N/A
 
Description

Description

Easily connect to Justrite safety containers using quick disconnects

Warranty

None

Specifications

Specifications

1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F)
Justrite Quick-Disconnect
Solvent Safety
Solvent-Compatible Containers
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'