BUCHI Vapor Duct for Rotavapor
Vapor Duct for use with the Rotavapor.
Supplier:  BUCHI 032341
 View more versions of this product
Catalog No. 05-422-904
Explore available promotions
Description
Description
  • For glass assembly V and C
  • Includes Combi-Clip
Specifications
Specifications
Vapor Duct with Combi-Clip
Rotavapor R-114/124
24/40
22 mm