Eppendorf™ Varispenser™ 2, Bottle-Top Dispenser
Varispenser™ 2, Bottle-Top Dispenser is ideal for dispensing aliquots of liquid from large supply bottles.
Supplier:  Eppendorf™ 4966000037
 View more versions of this product
Catalog No. 05-413-553
Explore available promotions
Description
Description
Varispenser™ 2 is ideal for dispensing aliquots of liquid from large supply bottles.
  • Standard GL 45 thread on all sizes for most common bottle threads
  • Safety valve balls in discharge valve prevents leakage when discharge tube is not mounted
  • Screwable air vent cap for easy assembly of drying tube or microfilter
  • Oval-shaped dispenser housing for easy transportation
  • PFA seal of the sliding piston prevents seizing up
  • Fixation slide on interior toothed track for rapid and accurate volume adjustment
  • High chemical resistance of all dispensing components
  • Telescopic aspirating tubes for variable adjustment to bottle height
  • Fully autoclavable without disassembly
  • User adjustment option
Specifications
Specifications
Analog Bottletop Dispenser with Adapters
1
1 to 10 mL
Adapters GL 25, GL 28/S 28, GL 32, GL 38, S 40, and telescopic intake tube (125 to 240 mm)