Eppendorf™ Varispenser™ 2x Bottle-Top Dispenser with Recirculation Valve
Ideal for dispensing aliquots of liquid from large supply bottles.
$1440.82 - $2853.42
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
05-413-557
|
Eppendorf™
4967000014 |
0.2 to 2.0 mL |
Each for $1,440.82
|
|
|||||
05-413-558
|
Eppendorf™
4967000022 |
0.5 to 5.0 mL |
Each for $1,440.82
|
|
|||||
05-413-559
|
Eppendorf™
4967000030 |
1.0 to 10 mL |
Each for $1,440.82
|
|
|||||
05-413-560
|
Eppendorf™
4967000049 |
2.5 to 25 mL |
Each for $1,883.47
|
|
|||||
05-413-561
|
Eppendorf™
4967000057* |
5.0 to 50 mL |
Each for $1,968.13
|
|
|||||
05-413-562
|
Eppendorf™
4967000065 |
10 to 100 mL |
Each for $2,853.42
|
|
|||||
Description
Ideal for dispensing aliquots of liquid from large supply bottles.
- Recirculation valve
- Standard GL 45 thread on all sizes for most common bottle threads
- Safety valve balls in discharge valve prevents leakage when discharge tube is not mounted
- Screwable air vent cap for easy assembly of drying tube or microfilter
- Oval-shaped dispenser housing for easy transportation
- PFA seal of the sliding piston prevents seizing up
- Fixation slide on interior toothed track for rapid and accurate volume adjustment
- High chemical resistance of all dispensing components
- Telescopic aspirating tubes for variable adjustment to bottle height
- Fully autoclavable without disassembly
- User adjustment option
Specifications
1 |