missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Vented Polyethylene Stopper for Cylindrical Dewar Flasks

Vented stopper to minimize evaporation from the Dewar

$125.21 - $125.21

Specifications

Color Gray
Material Polyethylene
For Use With (Application) Lid
View More Specs

Products 3
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
FB1019614
View Documents
Fisherbrand™
FB1019614
350 to 1000 mL Dewar Flask
Each for $125.21
 
FB1019612
View Documents
Fisherbrand™
FB1019612
1900 mL Dewar Flask
Each for $125.21
 
FB1019613
View Documents
Fisherbrand™
FB1019613
4300 mL Dewar Flask
Each for $125.21
 
Description

Description

  • Polyethylene
  • Vented
  • For Lab Grade Cylindrical Dewar Flasks
Specifications

Specifications

Gray
Polyethylene
Lid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'