MSA™ Ventilation Smoke Tube Kits
- Allows for controlled generation of visible smoke
- Determines velocity and flow patterns of heating, ventilating and air-conditioning systems
- Available with glass or plastic tubes with break-off tips
$464.64 - $514.49
Catalog Number | Mfr. No. | Description | Material | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Material | Price | Quantity | |||||
189993391
|
MSA™
458480 |
Box of twelve ventilation smoke tubes | Plastic |
Pack of 12 Each for $514.49
|
|
|||||
189993414
|
MSA™
458481 |
Ventilation Smoke Kit, including aspirator bulb, two rubber plugs, and six smoke-producing tubes contained in plastic carrying case | Glass |
Each for $464.64
|
|
|||||
Description
Use these kits to determine the velocity of slow-moving air currents and establish their direction and flow patterns. MSA™ Ventilation Smoke Tube Kits are ideal for use in commercial buildings and industrial processing plants as well as in shafts, mines and tunnels.
- Allows for controlled generation of visible smoke
- Determines velocity and flow patterns of heating, ventilating and air-conditioning systems
- Available with glass or plastic tubes with break-off tips
Specifications
Break-off |