missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ VeriShield™ 100 Series Behind-the-Neck Ear Muffs

VeriShield™ Behind-the-Neck Ear Muffs fit as a neckband to complement any other PPE or equipment the worker may be wearing or using.

$18.63 - $39.96

Specifications

Color Black
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Application) Construction, Manufacturing, Oil and Gas, Mining, Steel and Metals, Welding, Agriculture
Pairs per Package 1 Ea.
View More Specs

Products 3
Catalog Number Mfr. No. CSA Z94.2 Noise Reduction Rating (NRR) Price Quantity  
Catalog Number Mfr. No. CSA Z94.2 Noise Reduction Rating (NRR) Price Quantity  
19-085-532
View Documents
Honeywell™
1035112VS
Class B 22 dB
Each for $18.63
Case of 10 Each for $160.10
 
19-085-534
View Documents
Honeywell™
1035114VS
Class AL 25 dB
Each for $31.79
Case of 10 Each for $273.20
 
19-085-535
View Documents
Honeywell™
1035116VS
Class AL 28 dB
Each for $39.96
Case of 10 Each for $343.30
 
Description

Description

  • Fits around the neck from behind
  • Wide cup opening is ideal for workers with large ears, or who wear hearing aids
  • Lightweight cushioned headband
  • Memory foam ear cushions
  • Oil-resistant cup surface withstands heavy-work environments and frequent cleaning
  • Features Sound Management Technology (SMT) which improves employee safety by blocking harmful noise while allowing voices and signals to be heard more naturally
  • Designed for use with other PPE
Specifications

Specifications

Black
Black
1 Ea.
No
Behind-the-head
Yes
Construction, Manufacturing, Oil and Gas, Mining, Steel and Metals, Welding, Agriculture
VeriShield
ABS
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'