missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ VeriShield™ Dielectric Multi-Position Ear Muffs

VeriShield™ Dielectric Multi-Position Ear Muffs can be adjusted and worn in three different positions depending on comfort and application.

$30.19 - $37.94

Specifications

ANSI S3.1974 Yes
Cushion Color Black
For Use With (Application) Construction, Electrical, Manufacturing, Oil and Gas, Mining, Steel and Metals, Agriculture
Pairs per Package 1 Ea.
Product Type VeriShield
View More Specs

Products 2
Catalog Number Mfr. No. CSA Z94.2 Noise Reduction Rating (NRR) Price Quantity  
Catalog Number Mfr. No. CSA Z94.2 Noise Reduction Rating (NRR) Price Quantity  
19-085-539
View Documents
Honeywell™
1035186VS
Class A 25 dB
Each for $30.19
Case of 10 Each for $113.20
 
19-085-483
View Documents
Honeywell™
1035187VS
Class AL 30 dB
Each for $37.94
Case of 10 Each for $325.20
 
Description

Description

  • Multi-Position comfortably fits a wide range of head sizes
  • Wide cup opening is ideal for workers with large ears, or who wear hearing aids
  • Lightweight cushioned headband
  • Memory foam ear cushions
  • Oil-resistant cup surface withstands heavy-work environments and frequent cleaning
  • Features Sound Management Technology (SMT)
  • Available in mid or high level attenuation
Specifications

Specifications

Yes
Construction, Electrical, Manufacturing, Oil and Gas, Mining, Steel and Metals, Agriculture
VeriShield
Multi-position
Black
1 Ea.
Yes
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'