missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
VeriSpec™ Tungsten Standard for ICP 1000 ppm in 4% NH3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$425.06 - $1090.72
Chemical Identifiers
CAS | 12027-67-7 |
---|---|
Molecular Formula | H24Mo7N6O24 |
Molecular Weight (g/mol) | 1163.85 |
MDL Number | MFCD00064633 |
InChI Key | MISTUIVWPMGXFX-UHFFFAOYSA-T |
PubChem CID | 71306766 |
IUPAC Name | hexaammonium dodecaoxododecacyclo[7.5.1.1³,âµ.1³,¹³.1âµ,â·.1â·,â¹.0¹,â´.0¹,â¶.0¹,â¸.0²,¹³.0¹¹,¹â´.0¹¹,¹âµ]heptamolybdoxane-2,4,6,8,14,15-hexakis(ylium) |
SMILES | [NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O=[Mo]123(=O)O[Mo]45(=O)(=O)O[Mo]67(=O)(=O)O[Mo]89(=O)(=O)O[Mo]%10%11(=O)(=O)O[Mo]%12(=O)(=O)(O1)[O+]2[Mo]([O+]34)([O+]56)([O+]%10%12)([O+]78)[O+]9%11 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
RV010360100
|
Ricca Chemical Company
RV010360100N |
100 mL |
Each for $425.06
|
|
||||||
RV010360250
|
Ricca Chemical Company
RV010360250N |
250 mL |
Each for $762.94
|
|
||||||
RV010360500
|
Ricca Chemical Company
RV010360500N |
500 mL |
Each for $1,090.72
|
|
||||||
Description
- 1000 ppm in 4% NH3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
12027-67-7 | |
1163.85 | |
MISTUIVWPMGXFX-UHFFFAOYSA-T | |
hexaammonium dodecaoxododecacyclo[7.5.1.1³,âµ.1³,¹³.1âµ,â·.1â·,â¹.0¹,â´.0¹,â¶.0¹,â¸.0²,¹³.0¹¹,¹â´.0¹¹,¹âµ]heptamolybdoxane-2,4,6,8,14,15-hexakis(ylium) |
H24Mo7N6O24 | |
MFCD00064633 | |
71306766 | |
[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O=[Mo]123(=O)O[Mo]45(=O)(=O)O[Mo]67(=O)(=O)O[Mo]89(=O)(=O)O[Mo]%10%11(=O)(=O)O[Mo]%12(=O)(=O)(O1)[O+]2[Mo]([O+]34)([O+]56)([O+]%10%12)([O+]78)[O+]9%11 |
Specifications
12027-67-7,1336-21-6,7732-18-5 | |
100 mL | |
97.7466%,4.08%,0.1734% | |
MFCD00064633 | |
MISTUIVWPMGXFX-UHFFFAOYSA-T | |
[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O=[Mo]123(=O)O[Mo]45(=O)(=O)O[Mo]67(=O)(=O)O[Mo]89(=O)(=O)O[Mo]%10%11(=O)(=O)O[Mo]%12(=O)(=O)(O1)[O+]2[Mo]([O+]34)([O+]56)([O+]%10%12)([O+]78)[O+]9%11 | |
1163.85 | |
High Purity |
Natural LDPE Bottle | |
93.9134%,3.92%,0.1666% | |
H24Mo7N6O24 | |
1 | |
Liquid | |
hexaammonium dodecaoxododecacyclo[7.5.1.1³,âµ.1³,¹³.1âµ,â·.1â·,â¹.0¹,â´.0¹,â¶.0¹,â¸.0²,¹³.0¹¹,¹â´.0¹¹,¹âµ]heptamolybdoxane-2,4,6,8,14,15-hexakis(ylium) | |
71306766 |