BD Veritor™ InfoWiFi Module
Modular add-on; barcoded scanning of test identifiers, result download to Excel and wireless data reporting
Supplier: BD 445010
Alert:
Not included: BD Veritor™ Plus Analyzer (Cat. No. B256066)
Description
- BD Veritor™ Plus System add-on
- Enables barcode scanning, data download, and wireless transmission of results
- Only compatible with BD Veritor™ Plus Analyzers running firmware v5.50 or higher
- Barcode scanning:
- Collect info for each test including specimen ID, operator ID, and kit lot info and expiry
- If enabled, data will be stored with test type and result, date, time, and device information
- If printer attached to device, info will print
- Data download:
- Stores the last 3,500 tests on hard drive in a .csv file
- Hard drive can be unlocked via scanning USB Port Unlock sticker that comes with analyzer with BD Veritor™ InfoWiFi module installed
- Once unlocked, analyzer can be plugged into a computer and results can be viewed in Excel
- Wireless data transmission:
- When coupled BD Synapsys™ Informatics, analyzer can wirelessly transmit test data in real-time through the InfoWiFi module
Specifications
InfoWifi Module | |
For use with BD Veritor™ Plus Analyzer | |
2.5 x 2.75 x 1.5 in. |
Barcode scanner and wireless transmission add-on module | |
USB cable to connect an analyzer to a computer |