3M™ Versaflo™ PAPR System Filters and Cartridges
Compatible with powered air purifying respirators in the TR-600 Series
Supplier: 3M™ 7100027551
Description
- Filters and cartridges work with belt-mounted Versaflo PAPR units
- Recommended for use in healthcare, pharmaceutical, heavy industrial, manufacturing and/or general manufacturing environments
- NIOSH approved
- 3M Catalog# TR-6320N, TR-6350N, TR-6360N, TR-6510N, TR-6530N, TR-6710N-5, TR-6820N
Compliance
NIOSH approved.
Specifications
NIOSH | |
Combination HE Particulate/Formaldehyde | |
Pharmaceutical | |
TR-6350N |
Magenta/Olive | |
Cartridge | |
TR-600 PAPR | |
10/Pk. |