3M™ Versaflo™ Respiratory M-Series Helmet Assembly
3M™ Versaflo™ Respiratory M-Series Helmet Assembly includes a flame resistant shroud and Speedglas™ Welding Shield.
Supplier:  3M™ 7100168651
Catalog No. 19-070-936
Explore available promotions
Description
Description
  • Flame resistant shroud and helmet cover
  • 10% weight reduction when compared to L-905SG
  • Protection from welding arc (ANSI Z87) plus spark and splatter
  • Larger viewing area compared to L-905SG
  • Able to lift visors in non-hazardous areas to help facilitate communications
  • Head top can be used for most applications including arc gouging, plasma cutting, grinding, MIG, TIG, and GMAW welding
  • When used in conjunction with NIOSH approved air source, is designed to help protect the user from certain airborne contaminants, and certain head, eye, face, and welding hazards
  • Compatible with 3M™ Adflo, 3M™ Versaflo™ Powered Air Purifying Respirators, and 3M™ Supplied Air
  • Compatible with 9100XXi ADF with Natural Color Technology
  • 3M Catalog# M-407SG
Specifications
Specifications
ANSI Z89.1 Type I, Class G
Welding
Leather
Headgear
Standard
Black
3M™ Versaflo™ Respiratory M-Series Helmet Assembly, Nomex™ Shroud, Speedglas™ Welding Shield
M-407SG
1 Ea.
Clear