3M™ Versaflo™ Vortemp Heating Assembly, V-200
Used to heat incoming air by up to 50°F (28°C) on 3M™ Supplied Air Respiratory Systems that use 3M™ Headgear L-Series.
Supplier:  3M™ 7000002080
Catalog No. 19-038977
Explore available promotions
Description
Description
  • This assembly requires an airflow of 60-95 lbf/in² (413.69 Kpa - 655.00 Kpa)
  • User can adjust airflow and temperature
Specifications
Specifications
1 Ea.