missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
STERIS Vesta-Syde™ SQ Disinfectant
Quaternary ammonium disinfectant is an excellent fungicidal, non-sterile concentrate
$579.31 - $579.31
Specifications
Product Type | Quaternary Ammonium Disinfectant |
---|---|
Form | Concentrate |
Sterility | Non-sterile |
Appearance | Yellow |
Certifications/Compliance | Certificate of analysis |
Catalog Number | Mfr. No. | Volume (English) | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (English) | Price | Quantity | ||||||
19415139
|
STERIS
633508CA |
1 gal. |
Case of 4 Each for $579.31
|
|
||||||
Description
- Made specifically for use on hard non-porous surfaces
- At a use-dilution rate of 0.8% v/v (1:128), the solution can be applied manually or sprayed
- Fungicidal efficacy can support reduced sporicide use to control mold contamination
- Formulated with biodegradable surfactants that provide excellent cleaning ability
- Microbial control against bacteria, fungi, and viruses proven using AOAC and EN protocols
- Active ingredients notified according to the Biocide Product Directive and registered with the US EPA
- Manufactured in accordance with the cGMP with complete lot traceability
Specifications
Quaternary Ammonium Disinfectant | |
Non-sterile | |
Certificate of analysis | |
Microbial control against bacteria, fungi and viruses in cleanrooms |
Concentrate | |
Yellow | |
0.8% v/v (1:128) | |
Mops and wipers |