Learn More
Microsolv Technology Vials 10Ml 46 X 22.5 Crm Ctc C
Supplier: Microsolv Technology Corporation 95025CT10C
Vials, Headspace, Crimp Top, Glass. Clear, 10ml, 20mm Serum Finish, 22.5x46mm outer dimensions, Beveled Top and Rounded Bottom. CTC Compliant. MicroSolv Brand. 100/PK.