Corning™ Videodrop Sample Holder
This 2-piece part securly holds the Videodrop glass slide in one position during analysis.
Supplier:  Corning™ VD1000SH
Catalog No. 01-257-554
Explore available promotions
Description
Description
This 2-piece part securly holds the Videodrop glass slide in one position during analyisis. The sample holder is magnetic ensuring the blue LED light path maintains consistent throughout the experiement resulting in reproducibility from one recording to the next.
Qty./Pk: 1/Pk
Qty./Cs: 1/Ea
Brand: Corning™
Specifications
Specifications
Vessel Holder