Remco™ Vikan™ 10 in. Foam Blade Squeegee
Remco™ Vikan™ 10 in. Foam Blade Squeegee is ideal for drying surfaces after a sanitizing process or for pushing food from one area to another.
Supplier:  Remco™ 77515
 View more versions of this product
Catalog No. 19-830-221
Explore available promotions
Description
Description
  • Provides effective removal of both water and food debris from food preparation surfaces
  • Blade cartridge can be detached for easy cleaning or replacement
  • Replacement squeegee blade
  • Color-coded
Specifications
Specifications
FDA Compliant
Thread type Euro Hygienic
1 Ea.
3.1 in.
White
Polypropylene, Foam Rubber
Small