missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Remco™ Vikan™ 24 in. Swivel Neck Foam Blade Squeegee
Remco™ Vikan™ 24 in. Swivel Neck Foam Blade Squeegee is designed to allow the tool to reach around machinery and into tight spaces.
Supplier: Remco™ 77642
Description
- Highly effective on all types of floors
- The swivel neck feature allows flexibility and superior mobility in tight and difficult-to-reach spaces
- The cartridge system blades can be removed for easy cleaning and replaced when worn out
- Cost effective and economical
- Color-coded
Specifications
FDA Compliant | |
Thread type Euro Hygienic | |
1 Ea. | |
4.3 in. |
Green | |
Polypropylene, Foam Rubber | |
Large |