missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Remco™ Vikan™ 28 in. Single Blade Ultra Hygiene Bench Squeegee
Remco™ Vikan™ 28 in. Single Blade Ultra Hygiene Bench Squeegee is ideal for removing water or food debris from food contact surfaces.
Supplier: Remco™ 71703
Description
- One-piece construction
- Single-blade squeegee
- Ultra-hygienic design that minimizes areas for water and food soils to collect
- Effective in areas requiring the highest level of hygiene
- The soft and flexible rubber blade is ideal for removing water or food debris from floors and food contact surfaces
- Hygiene squeegees are a good foundation for any HACCP or food safety program
- A splash guard helps prevent water from reaching the dried surface
- Color-coded
Specifications
FDA Compliant | |
Thread type Euro Hygienic | |
1 Ea. | |
3.5 in. |
Blue | |
Polypropylene, Rubber | |
Large |