Remco™ Vikan™ Liquid Transporting Pail
For transporting cleaning chemicals and hot or cold ingredients.
Supplier: Remco™ 56869
Description
- With drip-free spout
- Hand-grip raised from base and stainless-steel handle
- Calibrated for variety of measuring units
- Flat side prevents spillage
- Stores using coordinating wall bracket
Specifications
Transporting Pail | |
3 gal. | |
transporting cleaning chemicals, hot or cold ingredients |
Polypropylene | |
Black | |
121°C |