Vileda Professional™ Double Bucket Chassis with Two 6 Gallon Bucket with Casters
Compact bucket systems for clean environments designed for effective cleaning. Vileda Professional™ Double Bucket Chassis with Two 6 Gallon Bucket with Casters is perfect for use with all Roll-O-Matic™ Mops.
Supplier: Vileda Professional™ 149624
Description
- Solid and stable polypropylene plastic buckets and bails are 100% autoclavable
- Unique electro-polished sieve (sold separately) with hole pattern, integrated bucket splash guard and handle support
- Two 6 gallon buckets
- Electro-polished stainless steel chassis
- Chassis features unique design to keep bucket upright
- Stainless steel autoclavable casters
- Fixed stainless steel screw/nut connection on chassis
Specifications
Autoclavable |