missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Vinblastine sulfate, 96.0-102.0%

Supplier:  Thermo Scientific™ 203450250

 View more versions of this product

Catalog No. AC203450250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Vinblastine sulfate
Authentic
Glass bottle
MFCD08706468,MFCD00082457,MFCD08706468
15, 10179
-21.7
Solubility in water: insoluble. Other solubilities: soluble in alcohols,acetone,ethyl acetate,,chloroform,practically insoluble in petroleum ether
OS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(C1)CCC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC
909.06
909.05
143-67-9
96.0 to 102% (HPLC)
C46H60N4O13S
25mg
−21.70° (21°C c=2,CH3OH)
vinblastine sulfate
KDQAABAKXDWYSZ-JKDPCDLQSA-N
methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(13S,15S,17S)-17-ethyl-17-hydroxy-13-(methoxycarbonyl)-1,11-diazatetracyclo[13.3.1.0⁴,¹².0⁵,¹⁰]nonadeca-4(12),5,7,9-tetraen-13-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate; sulfuric acid
131709225
96.0-102%

Chemical Identifiers

143-67-9
909.06
KDQAABAKXDWYSZ-JKDPCDLQSA-N
131709225
OS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(C1)CCC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC
C46H60N4O13S
MFCD08706468,MFCD00082457,MFCD08706468
vinblastine sulfate
methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(13S,15S,17S)-17-ethyl-17-hydroxy-13-(methoxycarbonyl)-1,11-diazatetracyclo[13.3.1.0⁴,¹².0⁵,¹⁰]nonadeca-4(12),5,7,9-tetraen-13-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate; sulfuric acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child if i

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

GHS Signal Word: Danger

EINECSNumber : 205-606-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only