missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vinyl acetate, 99%, stab. with 8-12ppm hydroquinone, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A162470F

 View more versions of this product

Catalog No. AAA162470F


Explore available promotions

Description

Description

Vinyl acetate is used in the preparation of polyvinyl acetate emulsions and polyvinyl alcohol. It acts as a precursor to polyvinyl acetate. It serves as an ingredient in furniture-glue. It is also used to prepare copolymers with other monomers such as ethylene, acrylic acid. Further, it is used to make adhesives, paints and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyl acetate is used in the preparation of polyvinyl acetate emulsions and polyvinyl alcohol. It acts as a precursor to polyvinyl acetate. It serves as an ingredient in furniture-glue. It is also used to prepare copolymers with other monomers such as ethylene, acrylic acid. Further, it is used to make adhesives, paints and plastics.

Solubility
Slightly miscible with water.

Notes
Light sensitive. Incompatible with acids, bases, oxidizing agents, peroxides, chlorosulfonic acid, ethylene imine, hydrochloric acid, oleum, nitric acid, sulfuric acid and 2-aminoethanol.
Specifications

Specifications

Vinyl acetate
108-05-4
0.932
−8°C (18°F)
C4H6O2
CH2=CHO2CCH3
2500 mL
1209327
vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
XTXRWKRVRITETP-UHFFFAOYSA-N
ethenyl acetate
7904
86.09
stabilized with 8 to 12ppm hydroquinone
-100°C
72°C to 73°C
Sweet
1.396
MFCD00008713
UN1301
14,9992
Slightly miscible with water.
CC(=O)OC=C
86.09
CHEBI:46916
99%

Chemical Identifiers

108-05-4
86.09
XTXRWKRVRITETP-UHFFFAOYSA-N
7904
ethenyl acetate
C4H6O2
MFCD00008713
vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
CHEBI:46916
CC(=O)OC=C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P271-P281-P303+P361+P353-P304+P340-P308+P313-P370+P378q-P501c

H225-H332-H335-H351

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: VINYL ACETATE, STABILIZED

EINECSNumber : 203-545-4

RTECSNumber : AK0875000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only